AS-DV Firmware Version 1.6.0 Release Notes
Firmware version 1.6.0 for the AS-DV Autosampler, released in April 2013, corrects issues in the previous version related to the omission of 0.5 mL vials in factory calibration, which could cause false detection. Users should upgrade from version 1.5.0 to 1.6.0.
Description:
A new version of firmware for the AS-DV was released in April, 2013; this is version 1.6.0. The revision changes described in Section B of this Release Note Document will describe any changes, bug fixes, enhancements, and any known bugs. This firmware version will supersede all previous firmware versions for this product.
Environment:
- AS-DV Firmware Version 1.6.0
- AS-DV Firmware Version 1.5.0
Attachment(s):
| File | Last Modified |
|---|---|
| AS-DV Firmware Version 1.6.0 Release Notes.PDF | January 07, 2022 |