AS-DV Simultaneous Injection Instructions
The AS-DV and AS40 Simultaneous Injection Kit enables one autosampler to deliver samples to two independent ion chromatography systems, increasing sample throughput. The kit includes necessary parts and detailed installation instructions. It also provides guidelines for configuring the systems in Chromeleon software, ensuring each device has a unique name. Routine maintenance involves checking for equal flow through both injection valve waste lines.
Description:
Instructions to set up an AS-DV with an IC to perform simultaneous injections are included in the attached document
Environment:
AS-DV
IC
Attachment(s):
| File | Last Modified |
|---|---|
| 031932-02_MANUAL-INSTAL-SIM_INJ-AS-DV_AS401.pdf | August 20, 2024 |