Thermo Fisher Scientific
AS-DV Autosampler Injection Interruption and Sample Skipping Issue - Chinese
Description: 
When the high backpressure and blockage occurs in some of the AS-DV sample delivery components, a noticeable injection sound and sample skipping will take place, resulting in sequence interruption. This basic troubleshooting video illustrates how to measure the backpressures of the needle tip, sample delivering tubing assembly, and injection valve rotor seals by connecting the running pump to each component temporarily. Following the instructions in this video, the operators can identify the blocked component and perform basic corrective actions such as backflushing the needle tip and rotor seals, cutting the blocked tubing off, and or optimizing the instrument method settings.
Environment:
  • AS-DV Autosampler
  • Ion Chromatography
  • Was this article helpful?