PSB-ICSP-1065-AXP-AXP-MS-MicroUSBDriver
In November 2016, it was identified that updated AXP and AXP-MS pumps shipped since February 2016 required a micro-USB driver not included in Chromeleon. The multi-segment USB-to-serial cable was missing from shipments until July 2016. Users must either obtain the missing cable or download the SSI driver to use the micro-USB cable. Detailed driver installation instructions are provided to ensure proper configuration and operation with Chromeleon.
Description:
Updated versions of the AXP and AXP-MS pumps began shipping in February 2016. These are based on the SSI MX Class pumps.
Environment:
AXP and AXP-MS Pump Cables and Driver
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1065-AXP-AXP-MS-MicroUSBDriver.PDF | December 15, 2021 |