PSB-ICSP-1066 - AXP and AXP-MS - FlowRateIssues
In November 2016, it was found that updated AXP and AXP-MS pumps shipped since February 2016 had firmware issues. These issues included incorrect flow rate reporting to Chromeleon and sporadic changes in flow rate, causing high pressure alarms. The resolution involves updating the pump firmware to version 2.03 using a specific procedure. This update resolves the flow rate display issue and prevents sporadic flow rate changes. Detailed instructions are provided for the firmware update process.
Description:
Updated versions of the AXP and AXP-MS pumps began shipping in February 2016. These are based on the SSI MX Class pumps.
Environment:
AXP and AXP-MS Pumps
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1066-AXP-AXP-MS-FlowRateIssues.PDF | December 15, 2021 |