Aquion Firmware 3.1.0 and Release Notes
A new firmware version 3.1.0 for the Aquion RFIC was released on June 20, 2019, addressing compatibility issues with Chromeleon 7.2.10 and future releases. This version supersedes all previous versions and is compatible with all Chromeleon 7 software and previous Aquion hardware products. Known bugs include Chromeleon 7.2.10 not recognizing Aquion modules with firmware 3.0.0 or earlier, resolved by upgrading to firmware 3.1.0 using Chromeleon 7.2.9 DUa.
Description:
The following information pertains to the Aquion and Aquion RFIC firmware version 3.1.0. This release supersedes the current firmware release due to a compatibility issue with Chromeleon 7.2.10 and future releases.
Environment:
Aquion and Aquion RFIC firmware version 3.1.0
Attachment(s):
| File | Last Modified |
|---|---|
| Aquion_3.1.0.fmw | March 09, 2022 |
| Aquion Firmware 3.1.0 Release Notes.PDF | December 13, 2021 |