Cindion Driver 1.1.29
Description:
To connect Cindion to Chromeleon (CM) a minimum CM version of 7.3.2 MuD is required. After installation of CM, the driver can be installed that will allow the Cindion to connect to the Instrument Controller.
Environment:
- Cindion
- Chromeleon
Procedure:
- Download the Cindion driver.
- To install the driver, double-click the file the "Cindion Driver Setup Wizard" should pop up. Follow the prompts to install the driver.
- In some cases, an error may trigger indicating that the file is not safe. Download the driver, a warning message may appear saying that the driver is not safe. To fix this issue, right click the download file and select "Keep". This issue occurs when Microsoft Edge browser is used. The preferred browser is Google Chrome to avoid this.
- This will open a new window. Select "Show More" and "Keep Anyway".
Attachment(s):
| File | Last Modified |
|---|---|
| clipboard_ef5696e84cf7b9f0cbd4e2695aea9a0a0.png | July 02, 2025 |
| clipboard_e9522d6523f0e34e3ba39ec4eabefb2f7.png | July 02, 2025 |
| clipboard_ebb556e716a0e45aafce86f253ac8eaf5.png | July 02, 2025 |
| clipboard_e38a6a28e96f7bd0c00b1f7c2d7992fd4.png | July 02, 2025 |
| Cindion Installer 1.1.29 Signed.msi | July 01, 2025 |