How to update Cindion firmware
Description:
This procedure describes how to update firmware for the Cindion.
Environment:
- Cindion
Procedure:
- Download the approriate Cindion firmware from the designated location.
- Connect a USB Type-B cable between the host PC and the Cindion SERVICE port located on the rear.
- Verify that a removable storage device appears on the host PC. The drive letter may vary.
- Copy the firmware (e.g. "update-1.1.42.zip") into the root directory on the removable storage device.
- Safely disconnect the USB cable the Cindion.
- Power cycle the system but pressing the power button on the top of the Cindion, then switch off the main power switch on the rear of the unit. Wait a few seconds, then turn the system back on using the rear power switch.
- Allow the system to complete its startup sequence. The firmware update will occur automatically during the startup.
Note: DO NOT INTERRUPT POWER DURING THE UPDATE PROCESS. - Verify the firmware update is completed by launching Chromeleon and navigating to the Cindion settings panel.
- Verify that the firmware version (LHC) matches the installed version. If the version does not match, repeat the firmware update procedure.
Attachment(s):
| File | Last Modified |
|---|---|
| clipboard_ebf5af3b903583a407359af86d4323b6e.png | April 08, 2026 |
| clipboard_e4600e1a5e6e47471e6a8ef0bdf67585e.png | April 08, 2026 |
| clipboard_ec80e7fa24450874e6627ed37bf41cbe2.png | April 08, 2026 |
| clipboard_ed8ecbc425df2c29149395de97ed9e951.png | April 08, 2026 |
| clipboard_eb53e35b6864e61ac15802cee2e08fc2b.png | April 08, 2026 |