PSB-IC-SYS-1056 - Integrion RFIC WiFi PCB
New Integrion RFIC systems and WiFi upgrade kits shipped before March 28, 2016, may have incorrectly configured WiFi PCBs. This could prevent the system from broadcasting a WiFi signal. If affected, check the S3 switch position on the WiFi PCB and adjust it to "RUN" or replace the PCB with part number 22153-61011R.
Description:
Integrion systems may have WiFi PCBs that are incorrectly configured.
Environment:
Integrion RFIC (Wi-Fi Enabled) and Integrion WiFi Upgrade Kits between 16021052 and 16040000.
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-IC-SYS-1056 - Integrion RFIC WiFi PCB.pdf | November 09, 2021 |