PSB-IC-SYS-1062 - Integrion ICS performing combustion IC applications
Integrion ICS systems performing combustion IC applications may experience inconsistent response to the TTL output signal from the combustion IC module, preventing data acquisition from starting. For resolution, contact ICSP Product Support by emailing ICSP.ProductSupport@thermofisher.com.
Description:
Integrion TTL input does not respond consistently to the TTL output signal from the combustion IC module.
Environment:
- Integrion ICS.
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-IC-SYS-1062 - Integrion ICS performing combustion IC applications.pdf | November 09, 2021 |