PSB-ICSP-1071 - RFID Causes Queue Interruptions
RFID Read/Write failures in Integrion HPIC systems can interrupt sample lists or injection queues. To resolve this, optimize RFID tag positions and adjust workflows to allow complete Read/Write cycles before starting the queue. A Chromeleon update is planned to mitigate manual adjustments. Ignore error codes #235 to #244 during the Read/Write process.
Description:
RFID Read/Write failures can cause the interruption of a sample list and/or an injection queue.
Environment:
All Integrion HPIC systems with RFID.
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1071 - RFID Causes Queue Interruptions.pdf | November 09, 2021 |