PSB-ICSP-1073 - Integrion and ICS-5000 Plus Pump CPU PCB
The bulletin addresses connectivity issues and potential blue screen errors in Integrion HPIC and ICS-5000+ DP/SP pump modules due to a faulty FTDI chip on the pump CPU PCB. It recommends power cycling and, if needed, replacing the affected PCBs with reworked versions containing the PDI chip. Contact Product Support if issues persist.
Description:
Pump connectivity issues and the pump driver not being recognized, nor installed properly. Isolated incidences of blue screen of death BSoD reported on PCs. The Supplier changed a chip on the Integrion pump CPU PCB as well as the ICS-5000+ mainboard PCB.
Environment:
- Integrion instruments manufactured from August to October 2017, S/Ns 1708XXXX through 1710XXXX
- For ICS-5000+ DP/SP pump modules manufactured from July to October 2017, S/Ns 1707XXXX through 1710XXXX
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1073 - Integrion and ICS-5000 Plus Pump CPU PCB.pdf | November 09, 2021 |