PSB-ICSP-1085 - Pinched RFID Wire
Internal reports indicate that a pinched RFID wire in Integrion systems with certain serial numbers could cause a short or electrical shock. Inspect the RFID Interface PCB for proper installation and replace any damaged wires. Ensure the wire is routed correctly to prevent pinching.
Description:
A pinched RFID wire that, if installed incorrectly, could cause a broken or exposed wire by being pinched between the system chassis and side panel.
Environment:
Integrion Systems: S/Ns: 1901xxxx through 1905xxxx
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1085 - Pinched RFID Wire.pdf | November 09, 2021 |