PSB-ICSP-1097 - Rev A - Conflict When Configuring Multiple Integrions On One Computer
Dionex Integrion HPIC systems may encounter configuration errors when multiple systems are connected to one computer due to non-unique MAC addresses. To resolve this, use separate computers for each system or replace the CPU PCB for one system if errors occur. Contact IC/SP PSE Group if issues persist.
Description:
Reports of Dionex Integrion HPICs having a configuration error when two or more Integrion systems are configured on one computer. Further, some Integrion CPU boards were not assigned unique MAC addresses at the factory. The factory has started to screen the affected instruments before shipping.
Environment:
Dionex Integrion Systems: S/N: 2001XXXX to 2012XXXX
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1097 - Rev A - Conflict When Configuring Multiple Integrions On One Computer.pdf | November 09, 2021 |