PSB-ICSP-1105 - Rev A - Integrion Communication Disconnects
Integrion HPIC systems are experiencing USB communication disconnects with Chromeleon 7.2 and 7.3 on various Windows OS and PC brands. An internal investigation is ongoing to identify a permanent solution. Recommendations include optimizing power settings, removing PCs from UPS or networks, and using high-quality USB cables or isolators.
Description:
For the most current approach, Please use knowledge article USB disconnect on Integrion.
There continue to be reports of the Integrion HPICs having communication disconnects. The Factory understands that these interruptions are adversely affecting our Customer’s productivity and our FSE’s efficiency with installations and repair.
The PSB provides a list of recommended troubleshooting steps. This list is for guidance only and one may have to perform multiple measures to resolve the issue(s).
Environment:
Integrion HPIC Systems
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1105 - Rev A - Integrion Communication Disconnects.pdf | January 21, 2022 |