Integrion - How to Download Firmware
To operate the Dionex DRS 600 Suppressor correctly, ensure your Dionex Integrion HPIC system firmware is version 1.7.0 or later. Earlier versions may cause compatibility errors. Download the latest firmware from the Unity Lab Services Knowledge Base, and upload it using the Chromeleon Instrument Configuration Manager.
Description:
This document explains how to sdownload firmware for an Integrion HPIC system.
Environment:
Integrion HPIC system
Attachment(s):
| File | Last Modified |
|---|---|
| Integrion - How to Download Firmware.pdf | November 21, 2021 |