Integrion Firmware 1.8.0 and Release Notes
A new firmware version 1.8.0 for the Integrion IC System was released on November 22, 2024, superseding all previous versions. This update resolves the issue of high-pressure valve counters resetting to 0 after power cycles. There are no enhancements or known bugs. Users should update to firmware version 1.8.0 for optimal performance.
Description:
Integrion Firmware 1.8.0
Release Date 16 December 2024
Environment:
Integrion HPIC
Attachment(s):
| File | Last Modified |
|---|---|
| Integrion Firmware v 1.8.0 Release Notes.pdf | December 17, 2024 |
| integrion Firmware 1.8.0.zip | December 10, 2024 |