Updating the Integrion HPIC Firmware for the DionexTM DRS 600 Suppressor
To update the Integrion HPIC firmware for the Dionex DRS 600 Suppressor, follow these steps: upgrade the firmware to version 1.7.0, reinitialize the suppressor modes in the Instrument Configuration Manager, and configure the suppressor modes in the Chromeleon Console. Ensure Chromeleon 7.2.7 or later is installed before starting.
Description:
This document explains how to successfully upgrade and configure an Integrion HPIC system to work with the new Dionex DRS 600 suppressor.
Environment:
Integrion HPIC system with the Dionex DRS 600 suppressor
Attachment(s):
| File | Last Modified |
|---|---|
| Updating the Integrion HPIC Firmware for the DionexTM DRS 600 Suppressor.pdf | November 21, 2021 |