Thermo Fisher Scientific
How to check BIOS Settings/Version
Issue
I am having disconnection issues with my instrument, where can I locate the BIOS settings on my computer to confirm that I have the latest version?
Environment
Integrion
Resolution
1. To bring up the Run dialog box, press Windows key + R key.
2. In the run dialog box, type regedit to access the Registry Editor.
3. Click on: HKEY_LOCAL_MACHINE\HARDWARE\DESCRIPTION\BIOS to see the BIOS settings and version. Refer to Figure 1
Registry Editor.png
Figure 1: Registry Editor 
  • Was this article helpful?