Pump will not start
Issue
How do I troubleshoot when pump will not start?
Environment
- Integrion models:
- 22153-60301
- 22153-60302
- 22153-60303
- 22153-60304
- 22153-60305
- 22153-60306
Resolution
Possible Causes and Solutions:
1. Possible Cause: Flow rate is set to zero.
Solution:
- Select a flow rate on the Chromeleon ePanel or the mobile app Home page.
- Verify that a flow rate is set in the Chromeleon instrument method.
2. Possible Cause: While being primed, pump starts briefly and an alarm occurs.
Solution:
- The high pressure limit was tripped.
- The low-pressure limit was tripped.
- A leak was detected.