155028 - Rev 01 - Dionex Easion Ion Chromatography System Operator Manual
Description:
The Thermo Scientific™ Dionex™ Easion™ Ion Chromatography System (Dionex Easion) performs isocratic ion analyses using suppressed conductivity detection. The Dionex Easion is an integrated ion chromatography system consisting of a pump, an injection valve, and a conductivity cell. Other system components (guard column, separator column, and suppressor) are ordered separately. The attached document describes the operation of this instrument.
Environment:
Thermo Scientific Dionex Easion Ion Chromatography System
Attachment(s):
| File | Last Modified |
|---|---|
| 155028 - Rev 01 - Dionex Easion Ion Chromatography System Operator Manual.pdf | October 09, 2025 |