Thermo Fisher Scientific
065289 - Rev 03 - Dionex ICS-1100 Ion Chromatography
Description: 
The Thermo Scientific Dionex™ ICS-1100 Ion Chromatography System (Dionex ICS-1100) performs ion analyses using suppressed or non-suppressed conductivity detection. An ion chromatography system typically consists of a liquid eluent, a high-pressure pump, a sample injector, a guard and separator column, a chemical suppressor, a conductivity cell, and a data collection system.
Environment:
Thermo Scientific Dionex™ ICS-1100 Ion Chromatography System (Dionex ICS-1100)