Thermo Fisher Scientific
031917 - Rev 02 - ICS-1500 Ion Chromatography System
Description: 
The Dionex ICS-1500 Ion Chromatography System ICS-1500 performs ion analyses using suppressed or non-suppressed conductivity detection. An ion chromatography system typically consists of a liquid eluent, a high-pressure pump, a sample injector, a guard and separator column, a chemical suppressor, a conductivity cell, and a data collection system.
Environment:
  • The Dionex ICS-1500 Ion Chromatography System
  • Was this article helpful?