Thermo Fisher Scientific
065291- Rev 03 - ICS-2100 - Dionex ICS-2100 Ion Chromatography System Operators Manual
Description: 
The Thermo Scientific Dionex ICS-2100 Ion Chromatography System Dionex ICS-2100 performs ion analyses using suppressed or non-suppressed conductivity detection. An ion chromatography system typically consists of a liquid eluent, a high-pressure pump, a sample injector, a guard and separator column, a chemical suppressor, a conductivity cell, and a data collection system.
Environment:
  • Thermo Scientific Dionex ICS-2100 Ion Chromatography System