031933 - Rev 1 - ICS-1000, ICS-1500, ICS-2000 Service Manual
The ICS 1000/1500/2000 Service Manual provides detailed service instructions. It covers system components, optional components, software requirements, and autosampler compatibility. The manual includes sections on theory of operations, repair and replacement procedures, calibration procedures, spare parts, and bills of materials. It emphasizes the importance of following proper procedures for maintenance and calibration to ensure system reliability and performance.
Description:
The ICS-1000, 1500, and 2000 are dedicated, isocratic ion chromatographs designed to run anion and cation applications with 3, 4, and 5 mm columns.
Environment:
- ICS-1000
- ICS-1500
- ICS-2000
Attachment(s):
| File | Last Modified |
|---|---|
| 031933 - Rev 1 - ICS-1000 ICS-1500 ICS-2000 Service Manual.PDF | December 16, 2021 |