PSB-SV-510 - ICS-3000 Series Variable Wavelength Detector
The ICS-3000 VWD firmware version 1.12.00 addresses deuterium lamp ignition failures. If the lamp fails to ignite, update the firmware and if necessary, replace the lamp power supply. This version improves ignition voltage and adds a wait period before re-ignition.
Description:
Deuterium D2 lamps sometimes fail to ignite for the ICS-3000 variable wavelength detector VWD. In some cases, when attempting to re-ignite a D2 lamp immediately after the lamp was switched off, e.g., after a power glitch, the lamp may fail to ignite. The associated error message is UV lamp ignition failure LD.
Environment:
- ICS-3000 Series Variable Wavelength Detector
- Assembly ID number 8010145
- Assembly ID number 8009744
- Assembly ID number 8010124
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-SV-510 - ICS-3000 Series Variable Wavelength Detector | June 07, 2025 |