065096 - Rev 1 - ICS-3000 Pump
The ICS-3000 Pump Service Manual provides detailed instructions for servicing the ICS-3000 pumps. It covers components such as pump heads, pressure transducers, piston seal wash systems, and vacuum degassing modules. The manual includes procedures for maintenance, repairs, troubleshooting, and calibration.
Description:
With flow rates ranging from 0.05 to 10.0 mL min and operating pressures up to 35 MPa 5000 psi, the DP/SP is designed for both standard and microbore applications. The pump’s patented isokinetic pre-compression phase allows a precise, almost pulse-free flow.
NOTE: The ICS-3000 has been discontinued and part number 062053 is no longer available to order. This may apply to other part numbers as well. Please check relevant PSBs for additional information.
Environment:
- ICS-3000 Dual Pump DP
- ICS-3000 Single Pump SP
Attachment(s):
| File | Last Modified |
|---|---|
| 065096 - Rev 1 - ICS-3000 Pump.PDF | June 07, 2025 |