PSB-ICSP-1073 - Integrion and ICS-5000 Plus Pump CPU PCB
Thermo Fisher Scientific has identified connectivity issues and potential blue screen errors with Integrion and ICS-5000+ DP/SP pump modules shipped between July and October 2017. The issues are due to a faulty FTDI chip on the pump CPU PCB. Affected PCBs should be replaced with ones containing the PDI chip. For unresolved issues, contact IC/SP Product Support Engineering.