ICS-3000, ICS-5000, ICS-5000 Plus DPSP FW 6.1.0 and Release Note
A new firmware version 6.1.0 for the ICS-3000/5000/5000+ DPSP was released on December 7, 2013, superseding previous versions. This update corrects a flow rate display issue and allows the ICS-5000+ DP/SP to operate up to 5000 PSI with the EGC 500. Users should update to the latest firmware via the Chromeleon Device Driver.
Description:
Firmware release 6.1.0. The attached firmware applies to all listed instruments.
Environment:
- ICS-3000
- ICS-5000
- ICS-5000 Plus
- ICS-5000 DP
- ICS-5000 SP
Attachment(s):
| File | Last Modified |
|---|---|
| dpsp3000_6.1.0.hex | March 10, 2022 |
| ICS-3000, ICS-5000, ICS-5000 DPSP - FW 6.1.0 - Release Note - 20140206 | December 30, 2021 |