075836 - Rev 1 - ICS-5000 DPSP - Service Manual - Apr 2011
The ICS-5000 Pump Service Manual outlines maintenance and repair procedures for the ICS-5000 Dual and Single Pumps (DP, SP). It includes details on replacing components such as check valves, piston seals, and vacuum degassing chambers. The manual emphasizes using original Dionex parts and following safety precautions. It also provides instructions for priming the pump, calibrating system pressure, and accessing the electronics compartment.
Description:
Each ICS-5000 pump can be configured for either capillary IC applications or analytical (standard bore and microbore) IC applications. Capillary IC pumps are always isocratic (they deliver one eluent).
Environment:
- ICS-5000 Dual Pump DP
- ICS-5000 Single Pump SP
Attachment(s):
| File | Last Modified |
|---|---|
| 075836 - Rev 1 - ICS-5000 DPSP - Service Manual - Apr 2011.PDF | December 16, 2021 |