Thermo Fisher Scientific
ICS-6000 Letter of Equivalency
The Thermo Scientific Dionex ICS-6000 HPIC system, introduced in 2018, replaces the Dionex ICS-5000+ HPIC system. It maintains the same fit, form, and function while adding features like a Consumables Device Monitor, support for tablet-based interface with Chromeleon CDS software, and Dual Eluent Generator Cartridge mode for complex carbohydrate analysis. It uses the same columns and Eluent Generation Cartridges, ensuring equivalent chromatographic performance.
Description: 
ICS-6000 LOE Customer Letter
Environment:
ICS-6000