065759 - Rev 01 - Dionex ICS-6000 Consumables Tracking Quick Start Guide
The Thermo Scientific Dionex ICS-6000 Ion Chromatography System uses RFID-enabled consumables tracked by the Consumable Device Monitor (CDM). Connected to the DC and configured in Chromeleon 7, the CDM identifies and tracks consumable usage. Setup involves connecting the CDM via USB, configuring it in Chromeleon, ensuring correct RFID tag placement, scanning consumables, and selecting them for tracking. Before removing a consumable, data is written to its RFID tag.
Description:
The attached guide provides information for operating the Consumable Device Monitor (CDM) a smart RFID controller that provides the necessary interface to
the consumables used with the Thermo Scientific™ Dionex™ ICS-6000 Ion Chromatography System that are capable of RFID (radio-frequency identification) communication.
Environment:
ICS-6000
Attachment(s):
| File | Last Modified |
|---|---|
| 065759 - Rev 01 - Dionex ICS-6000 Consumables Tracking Quick Start Guide.pdf | October 09, 2025 |