Thermo Fisher Scientific
60-065483 - Rev C - Dionex ICS-5000 Plus, ICS-6000 Dual Pump and Single Pump Preventive Maintenance Procedure
The Dionex ICS-5000+/ICS-6000 preventive maintenance involves replacing parts like piston seals, O-rings, and check valves. Recommended every 18 months for capillary pumps and annually for analytical pumps. Each pump takes 60 minutes, requiring specific tools and kits.
The Thermo Scientific Dionex ICS-5000+/ICS-6000 Dual and Single Pump Preventive Maintenance Procedure outlines the steps for replacing key parts in each pump, including main piston seals, check valves, and tubing. Preventive maintenance is recommended every 18 months for capillary pumps and annually for analytical pumps. The procedure includes a detailed list of required parts, tools, and instructions for disassembling, cleaning, and reassembling pump components to ensure optimal performance.
Description: 
This manual describes a preventive maintenance procedure for the products listed in the Environment.
During this procedure, the following parts in each pump are replaced:
  • Main piston seals
  • Piston seal wash body O-rings
  • Piston seal wash seals
  • Inlet and outlet check valves (primary pump head only)
  • Priming valve seal (secondary pump head only)
  • Piston seal wash system tubing
Thermo Fisher Scientific recommends performing preventive maintenance every 18 months for capillary pumps and annually for analytical pumps.
Environment:
  • Thermo Scientific™ Dionex™ ICS-5000+ Dual Pump (DP)
  • Thermo Scientific™ Dionex™ ICS-5000+ Single Pump (SP)
  • Thermo Scientific™ Dionex™ ICS-6000 Dual Pump (DP)
  • Thermo Scientific™ Dionex™ ICS-6000 Single Pump (SP)