Rev 1 - ICS-6000 EG - Service Manual
The ICS-6000 Pump Service Manual provides detailed maintenance instructions, including accessing electronics, replacing tubing, calibrating pressure, and replacing components like valves and degassing chambers. It emphasizes safety and using Thermo Fisher parts.
Description:
The ICS-6000 EG generates high-purity acid or base eluents online from deionized water.The EG can be configured for single- or dual-channel operation.Each channel includes a high precision programmable current source (power supply).
Environment:
- ICS-6000 EG
Attachment(s):
| File | Last Modified |
|---|---|
| Rev 1 - ICS-6000 EG - Service Manual.PDF | December 16, 2021 |