PSB-ICSP-1076 - 2L Bottle Leakage
The 2L bottles (P/N 062510) manufactured between July 11, 2018, and September 10, 2018, were recalled due to air leakage issues caused by irregular sealing surfaces. Bottles shipped after September 11, 2018, are properly manufactured. If issues persist, replace the bottle under warranty.
Description:
Two liter bottle P/N 062510 (ASSY, BTL, 2L, ELUENT) was manufactured incorrectly. All bottles in distribution centers, received from San Jose between 7-11-2018 and 9-10-18, were recalled to the factory to be replaced by good product. All bottles shipped to service engineers or customers from the factory or distribution centers after September 11, 2018 will be properly manufactured and thoroughly inspected.
Environment:
IC systems
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1076 - 2L Bottle Leakage.pdf | November 16, 2021 |