PSB-ICSP-1087 ERD 500 Adaptor Cable
Thermo Fisher Scientific advises customers using a Dionex ERD 500 Electrolytically Regenerated Desalter in an ICS-6000 system to use an adaptor cable due to Chromeleon driver incompatibility. The driver fix will be in CM7.3 release. Contact IC/SP Ion Chromatography Product Support for the adaptor cable.
Description:
This PSB is to be applied only to customers who need to use a Dionex ERD 500 Electrolytically Regenerated Desalter in an ICS-6000 system. Currently, the Chromeleon driver for the ICS6000 does not support ERD 500 desalter 1-wire function in CM 7.2.10 and previous versions.
Environment:
- Dionex ERD 500 Electrolytically Regenerated Desalter
- Chromeleon
- ICS-6000
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1087 ERD 500 Adaptor Cable.PDF | December 14, 2021 |