PSB-ICSP-1096 - Sodium Acetate Salt Notification
Reports of color variance in Electrochemical-grade Sodium Acetate Salt (P/N 059326) do not affect its performance for HPAE-PAD methods. The factory has tested all lots for quality. It is recommended to filter the sodium acetate solution using Nalgene Rapid-Flow Sterile Disposable Filter Units.
Description:
We have received reports of lot to lot color variances associated with Electrochemical-grade, Sodium Acetate Salt (Pre-weighed Reagent, P/N 059326). Please note that color variance of the residuals on the filter post filtration does not indicate a lowered performance of the sodium acetate eluent in the determination of carbohydrates and amino acids using HPAE-PAD methods.
Environment:
Sodium Acetate Salt (Pre-weighed Reagent, P/N 059326)
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1096 - Sodium Acetate Salt Notification.PDF | December 14, 2021 |