PSB-ICSP-1197 2mm IC Column End Fitting Improvement
Description:
The current 2 mm End Fitting (043278) PEEK plastic hardware used in 2 mm column applications has historically been prone to intermittent cracking during the column assembly process. Mitigation efforts with molding vendors over the years have had limited success, and the cracking issue still persists in some cases. Further investigation has determined that the existing part design is inadequate to meet the assembly torque and pressure specifications required for this application, thus has been re-design for better robustness.
The recommended design revision increases wall thickness at the identified high-stress region, improving torque and operating pressure capability by approximately 2.5–3×. This change is intended to eliminate end-fitting leakage and reduce the incidence of leaking columns. The redesigned 2 mm column end fittings are scheduled to begin shipping from 11th March 2026.
Environment:
2mm Ion Chromatography Columns
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1197 2mm IC Column End Fitting Improvement.pdf | April 02, 2026 |
| IC Column End-fitting Improvement Customer Letter Feb 2026.pdf | April 02, 2026 |