Thermo Fisher Scientific
High Internal Backpressures and Leaking Issues of the Suppressors - Chinese
Description: 
This troubleshooting video illustrates the most common suppressor issues: high internal backpressure, external leaking and internal leaking. Backflushing the suppressors could possibly remove the contaminants causing internal channel blockage. After watching this video, operators will be able to distinguish the external leaking and internal leaking issues, backflush the suppressor, and hydrate and install a new replacement suppressor to return the system to normal operation.
Environment:
  • Ion Chromatography
  • Suppressors
  • Was this article helpful?