ICS-Series VWD - Firmware 1.60 and Release Notes
A new firmware version 1.60 for ICS-Series VWD was released on June 1, 2020, superseding all previous versions. The firmware supports the USB controller chip FT120 and is compatible with all ICS-3000-VWD detectors and CM versions 6.80 SP1 and 7.0. Firmware updates are required to ensure compatibility and reliable operation, especially before any main board exchanges. Firmware 1.60 prevents downgrading to older versions to maintain hardware compatibility.
Description:
This release is the required to support the new USB controller chip FT120 (new main board product number 9074.0111). This new controller is a replacement for the obsolescent PDIUSBD12 USB controller.
Environment:
ICS-Series VWD
Attachment(s):
| File | Last Modified |
|---|---|
| ICS_VWD3000_1.60.hex | March 24, 2022 |
| ICS-Series VWD FW 1.6 release note.pdf | March 01, 2022 |