PSB-IC-SYS-1008 - ICS-3-5-SPDP-SVCBLOCK
Description:
After July 2011, ICS-5000 SP/DP pump module motors were upgraded with a new brushless motor design to increase pump reliability. The new pump mechanism is backwards compatible for both ICS-5000 and ICS-3000 SP/DP modules. The ICS-3000 Pump Service Manual (Document No. 065096) and ICS-5000 Pump Service Manuals (Document 075836) will be updated to include additional information for installing the latest version (Rev. 07) of the SP/DP Mechanical Block (062060 PUMP DRIVE
ASSY,SVC,ICS3/5).
Environment:
- ICS-3000
- ICS-5000
- ICS-3000 SP/DP
- ICS-5000 SP/DP
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-IC-SYS-1008-ICS-3-5-SPDP-SVCBLOCK.PDF | March 16, 2022 |