PSB-ICSP-1101 - Rev A - Easion CD Calibration
Recent reports indicate implausible conductivity values in Dionex Easion IC systems due to an incomplete conductivity offset calibration step. The factory corrected this in May 2021. If negative or high conductivity signals are observed, recalibration of the conductivity cell offset and slope is required.
Description:
Recent reports of implausible conductivity values have been observed in Dionex Easion Ion Chromatography systems. An incomplete conductivity offset calibration step was found in the Factory Operational Qualification (FOQ) and calibration process.
As of May 2021, the conductivity calibration process has been performed by the factory per the FOQ.
Environment:
- Easion
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1101 - Rev A - Easion CD Calibration.pdf | January 17, 2022 |