PSB-ICSP-1102 - Rev A - Pump Assembly Repair Procedure
The Dionex Aquion, Easion, and ICS-1100/1600/2100/600/900 pump assemblies can fail due to premature wear. Instead of replacing the entire pump assembly, a repair kit (P/N 22176-62004) is available for field repair. If the repair does not resolve the issue, replace the entire pump assembly.
Description:
Pump assembly (P/N 058091T) on the Dionex Aquion and Easion, and ICS-1100/1600/2100/600/900 instruments can fail due to premature wear on the bearings and/or CAM. Instead of replacing the whole pump assembly, a pump repair kit, P/N 22176-62004, was created to allow repair in the field.
Environment:
- ICS-1100
- ICS-1600
- ICS-2100
- ICS-600
- ICS-900
- Aquion
- Easion
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1102 Rev A - Pump Assembly Repair Procedure | June 15, 2022 |