PSB-ICSP-1113 - Rev A - EGC 500 KOH and MSA Improvements
Description:
There have been multiple reports of EGC 500 KOH and MSA cartridges leaking and damaged upon receipt. The damaged cartridges have impacted installations, customer downtime, and exacerbated already strained supply channels. Resolving this problem as quickly as possible is of the utmost importance to the IC/SP Business Unit.
An investigation into the root cause of the leaks and subsequent damage was began in early 2021. After significant evaluation and testing, multiple root causes were found and included: poor seal between the bottle and pod, gap between surfaces/seals, and an O-ring issue. As a result of this investigation, the following improvements were implemented:
- Optimized O-ring, December 29, 2021
- Improved surface finish PODs, December 29, 2021
- Cover changed from Xenoy to Polypropylene for improved chemical resistance, December 21, 2021
Note: The change from Xenoy to polypropylene on the bottom cover was to reduce the chance of damage once a leak had occurred. It was not a cause of the leaks and does not affect performance of the cartridges.
There is not a visual difference between older EGCs and those with the above improvements so a S/N cut-in has been provided to help identify if a cartridge was manufactured before or after the improvements.
Environment:
P/N 075778 Dionex EGC 500 KOH Cartridge, S/N 211282201313 and later
P/N 075779 Dionex EGC 500 MSA Cartridge, S/N 211282911328 and later
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1113 - Rev A - EGC 500 KOH and MSA Improvements.pdf | February 25, 2022 |