PSB-ICSP-1137-Rev A-Changes to CV Mode for Suppressors
Description:
Constant Voltage (CV) or Dynamic mode was introduced with the DRS suppressors to offer an alternate approach to suppression where the voltage is held constant, and the current is allowed to fluctuate. CV mode has experienced a low adoption rate and proved beneficial in a limited number of applications. As a result, CV mode will no longer be actively promoted or marketed, and all references will be removed from customer facing material at the beginning of February 2024.
Environment:
Dionex Suppressors
Recommendations:
For customers that are successfully using CV mode and for those using Constant Current (CC) or Legacy mode no action is required. For customers that are experiencing problems with CV mode and for any new installations the recommendation is that only CC or Legacy mode be utilized.
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1137-Changes to CV Mode for Suppressors.pdf | January 24, 2024 |