PSB-ICSP-1157 Temporary Tubing Change in Inuvion Shipkit and ICS-6000 EG
PSB-ICSP-1157 addresses a temporary tubing change in Inuvion and ICS-6000 EG systems due to a material shortage from mid-November to mid-December 2024. Gray polyurethane tubing was used instead of clear tubing (P/N: 047203). Affected systems include Inuvion S/N 241151061 to 241151076 and ICS-6000 EG S/N 24116812 to 24116834. It is recommended to replace the gray tubing with clear tubing by ordering the appropriate lengths. For issues, contact IC/SP PSE Group.
Description:
Due to a material shortage of clear polyurethane tubing (P/N: 047203: TUBING,AIR,.062X.125 PU,CLR) from mid-November to mid-December 2024, the factory has shipped new ICS-6000 EG modules and Inuvion systems with an alternative tubing of the same size but in gray. This gray polyurethane tubing is used in the ICS-6000 EG Degas vent tubing kit, Inuvion pump waste tubing kit, and the spare waste tubing in the Inuvion shipkit during this period.
Environment:
ICS-6000 EG
Inuvion
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1157 Temporary Tubing Change in Inuvion Shipkit and ICS-6000 EG.pdf | January 21, 2025 |