PSB-ICSP-1173-AS-AP well plate and cover
Description:
We have received reports that the 2ml 96 deep well plate AS-AP tray PN: 066334 WELL-PLATE,96-WELLSINUIN,2.0ML,PP,5 PACK did not fit the recommended and documented cover PN: 066330 WELL-CAPS,96 SQUARE,2.0ML,SILICONE,10 PK. It was also noticed that there was other well plate trays listed which were not suitable for IC applications. So, a full investigation was conducted into all the tray and cover types.
Affected Well Plates and Covers:
· PN: 066330 WELL-CAPS,96 SQUARE,2.0ML,SILICONE,10 PK
· PN: 066332 WELL-PLATE,96-WELL,0.5ML,PP,10 PK
· PN: 066333 WELL-PLATE,96 WELLS V,0.45ML,PP,20PK
· PN: 066334 WELL-PLATE,96-WELLSINUIN,2.0ML,PP,5 PACK
· PN: 066335 WELL-CAPS,96-ROUND,0.5ML,SILICONE,10 PK
Environment:
AS-AP Autosampler
Recommendations:
During the investigation it was found that the correct cover PN:066335 WELL-CAPS,96-ROUND,0.5ML,SILICONE,10 PK fitted PN:066334 WELL-CAPS,96 SQUARE,2.0ML,SILICONE,10 PK. This was tested and verified to ensure full compatibility for the AS-AP sampler with PN:074939 SAMPLE TABLE, LOW- AS-AP. The label description and quantity per pack has also been updated.
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1173-ASAP well plate and cover.pdf | July 28, 2025 |