PSB-ICSP-1189 Defective Integrion PM Kit P/N 22153-62041
Description:
Due to field reports of incorrect valve rebuild kits inside of Integrion Preventive Maintenance kits, global containment was started on January 9, 2026. Further, there were reports of the incorrect HPLC rotor seal being received instead of the IC rotor seal. Containment and recall on P/N 22153-62041 (ASSY,KIT,PM,INTEGRION) where the P/N of 075973 (KIT,REBUILD, VLV, 6-PORT,6K) was IDEX P/N 7145-999 instead of 9915-999 in the PM kit was started. The subcomponents of the Integrion PM kit P/N 075973 were inspected at the factory and found to be correct, however, there is still a risk of incorrect PM kits. The factory was unable to inspect the Integrion PM kit as none were in stock, thus there may be further kits with issues.
Environment:
Integrion
PM kit
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1189 Defective Integrion PM Kit PN 22153-62041.pdf | January 16, 2026 |