PSB-ICSP-1201-Inuvion New Version Pump Seals
Description:
A. Background
To support dual sourcing/supply continuity, a new version of the main pump seal, P/N B51000563 (SEAL, ENERGIZER, FLANGLESS, 1/8 ID), has been validated and implemented for the Inuvion system.
The new pump seal has a different appearance from the previous version but is fully compatible with the Inuvion pump head. The installation procedure and pump performance remain unchanged.
Environment:
Inuvion
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1201-Inuvion New Version Pump Seal.pdf | July 01, 2026 |