PSB-ICSP-1202 Inspection of Cindion Solids Boat PN B51006443 for Conformity
Description:
Thermo Fisher Scientific has identified a supplier-related quality issue affecting some Cindion Solids Boats, P/N B51006443.
The issue results in deformation of the solids boat geometry, causing misalignment between the solids boat, transfer arm, and magnet assembly during operation.
Because the solids boat is not serialized, Thermo Fisher Scientific cannot identify specific affected units, manufacturing dates, or shipment ranges. Identification of nonconforming parts requires physical inspection using the procedure described in this bulletin.
The supplier investigation remains ongoing.
Environment:
- Cindion
- Combustion IC
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1202 Inspection of Cindion Solids Boat PN B51006443 for Conformity.pdf | July 17, 2026 |